(2R,3R)-(-)-2,3-Butanediol 98%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
D(-)-2,3-Butanediol, (2R,3R)-Butane-2,3-diol, (R,R)-(-)-2,3-Butylene glykol, (R,R)-(-)-2,3-Butanediol, (2R,3R)-(-)-2,3-Dihydroxy butane, (R,R)-(-)-2,3-Dihydroxy butane, (2R,3R)-(-)-2,3-Butylene glykol
Purity: 98%
CAS No.: 24347-58-8
Molecular Formula: C4H10O2
Formula Weight: 90.12
Flash Point: 85°(185°F)
Refractive Index: 1.4315
Melting Point: 16°
Boiling Point: 179-180°
Beilstein Reference: 4290593
Merck Reference: 14,1568
Optical Rotation: -13° (neat)
MDL No.: MFCD00064267
Formula:
C₄H₁₀O₂ Boiling Pt: 179…180 °C Melting Pt: 16 °C Density: 0.988 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064267 CAS Number: 24347-58-8 EINECS: 246-186-9 Merck Index: 14,1568 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...