6-Nitro-o-toluic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

L06563-06 L06563-14
CAAAAL06563-03EA 38.82 CAD
CAAAAL06563-03 CAAAAL06563-06 CAAAAL06563-14
6-Nitro-o-toluic acid 98%
6-Nitro-o-toluic acid

Purity: 98% CAS No.: 13506-76-8 Molecular Formula: C8H7NO4 Formula Weight: 181.15 Melting Point: 153-156° Beilstein Reference: 2050804

Formula: C₈H₇NO₄
Boiling Pt: 353 °C (1013 hPa)
Melting Pt: 153…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00007267
CAS Number: 13506-76-8
EINECS: 236-833-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR