(+)-Dimethyl-L-tartrate 99%

Supplier: Thermo Scientific Chemicals

L06561-14 L06561-22
CAAAAL06561-14EA 54.35 CAD
CAAAAL06561-14 CAAAAL06561-22
(+)-Dimethyl-L-tartrate 99%
(+)-Dimethyl L-tartrate

Purity: 99% CAS No.: 608-68-4 Molecular Formula: C6H10O6 Formula Weight: 178.14 Flash Point: 170°(338°F) Melting Point: 56-61° Boiling Point: 158°/11mm Beilstein Reference: 1726256 Optical Rotation: +21° (c=1 in water) MDL No.: MFCD00064437

Formula: C₆H₁₀O₆
Boiling Pt: 158 °C/11 mm
Melting Pt: 56…61 °C
Density: 1.30
Flash Pt: >170 °C(338 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064437
CAS Number: 608-68-4
EINECS: 210-166-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR