3-Amino-α-methylbenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: m-Amino-α-methylbenzyl alcohol, 1-(3-Aminophenyl)ethanol, 3-Aminophenyl methyl carbinol, 3-(1-Hydroxyethyl)aniline

L06513-14 L06513-22
CAAAAL06513-14EA 119.29 CAD
CAAAAL06513-14 CAAAAL06513-22
3-Amino-α-methylbenzyl alcohol 98%
3-Amino-α-methylbenzyl alcohol

Purity: 98% CAS No.: 2454-37-7 Molecular Formula: C8H11NO Formula Weight: 137.18 Flash Point: 157°(314°F) Melting Point: 66-69° Boiling Point: 217° Beilstein Reference: 2802415 MDL No.: MFCD00007798

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 217 °C
Melting Pt: 66…69 °C
Flash Pt: 157 °C (314 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007798
CAS Number: 2454-37-7
EINECS: 219-525-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR