Cyclohexanemethylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L06501-06
L06501-14
CAAAAL06501-06EA
63.36
CAD
CAAAAL06501-06
CAAAAL06501-14
Cyclohexanemethylamine 98%
Cyclohexanemethylamine
Purity: 98%
CAS No.: 3218-02-8
Molecular Formula: C7H15N
Formula Weight: 113.20
Flash Point: 43°(109°F)
Refractive Index: 1.4620
Melting Point: -8°
Boiling Point: 161-163°
Beilstein Reference: 635751
UN No.: 2734
MDL No.: MFCD00001520
Formula:
C₇H₁₅N Boiling Pt: 161…163 °C Melting Pt: –8 °C Density: 0.871 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001520 CAS Number: 3218-02-8 EINECS: 221-741-8 UN: 2734 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...