3',4',5'-Trimethoxyacetophenone 99%
Supplier: Thermo Scientific Chemicals
L06439-06
L06439-14
CAAAAL06439-06EA
73.63
CAD
CAAAAL06439-06
CAAAAL06439-14
3',4',5'-Trimethoxyacetophenone 99%
3',4',5'-Trimethoxyacetophenone
Purity: 99%
CAS No.: 1136-86-3
Molecular Formula: C11H14O4
Formula Weight: 210.23
Melting Point: 79-83°
Boiling Point: 173-174°/10mm
MDL No.: MFCD00008738
Formula:
C₁₁H₁₄O₄ Boiling Pt: 173…174 °C/10 mm Melting Pt: 79…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008738 CAS Number: 1136-86-3 EINECS: 214-501-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...