Octyl acetate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetic acid octyl ester

L06405-18 L06405-30
CAAAAL06405-18EA 46.77 CAD
CAAAAL06405-18 CAAAAL06405-30
Octyl acetate 98+%
Octyl acetate

Purity: 98+% CAS No.: 112-14-1 Molecular Formula: C10H20O2 Formula Weight: 172.27 Flash Point: 86°(186°F) Refractive Index: 1.4190 Boiling Point: 210-211° Beilstein Reference: 1754554 MDL No.: MFCD00009562

Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
Boiling Pt: 210…211 °C
Melting Pt: –38.5 °C
Density: 0.868
Flash Pt: 86 °C(186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009562
CAS Number: 112-14-1
EINECS: 203-939-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR