1,7-Heptanediol 98%
Supplier: Thermo Scientific Chemicals
L06384-06
L06384-14
CAAAAL06384-06EA
103.96
CAD
CAAAAL06384-06
CAAAAL06384-14
1,7-Heptanediol 98%
1,7-Heptanediol
Purity: 98%
CAS No.: 629-30-1
Molecular Formula: HO(CH2)7OH
Formula Weight: 132.20
Flash Point: 127°(260°F)
Refractive Index: 1.4550
Melting Point: 17-18°
Boiling Point: 258-260°
Beilstein Reference: 1633482
MDL No.: MFCD00002987
Formula:
C₇H₁₆O₂ Boiling Pt: 258…260 °C Melting Pt: 17…18 °C Density: 0.953 Flash Pt: 127 °C (260 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002987 CAS Number: 629-30-1 EINECS: 211-085-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...