cis-4-Decenal 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06382-03
L06382-06
CAAAAL06382-03EA
47.72
CAD
CAAAAL06382-03
CAAAAL06382-06
cis-4-Decenal 95%
cis-4-Decenal
Purity: 95%
CAS No.: 21662-09-9
Molecular Formula: C10H18O
Formula Weight: 154.25
Storage Temperature: 2°C to 8°C
Flash Point: 81°(177°F)
Refractive Index: 1.4440
Boiling Point: 78-80°/10mm
Beilstein Reference: 2323646
MDL No.: MFCD00007024
Formula:
C₁₀H₁₈O Boiling Pt: 78…80 °C/10 mm Density: 0.857 Flash Pt: 81 °C(177 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007024 CAS Number: 21662-09-9 EINECS: 244-514-5 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...