Trifluoroacetic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
TFA
L06374-AC
L06374-AE
L06374-AP
CAAAAL06374-ACEA
66.52
CAD
CAAAAL06374-AC
CAAAAL06374-AE
CAAAAL06374-AP
Trifluoroacetic acid 99%
Trifluoroacetic acid
Purity: 99%
CAS No.: 76-05-1
Molecular Formula: C2HF3O2
Formula Weight: 114.02
Refractive Index: 1.2840
Odor: Pungent
Boiling Point: 72-73°
Beilstein Reference: 742035
Merck Reference: 14,9681
UN No.: 2699
MDL No.: MFCD00004169
Formula:
C₂HF₃O₂ Boiling Pt: 72…73 °C Melting Pt: –15 °C Density: 1.480 Storage Temperature: Ambient |
MDL Number:
MFCD00004169 CAS Number: 76-05-1 EINECS: 200-929-3 UN: 2699 ADR: 8,I Merck Index: 14,9681 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...