Octadecyltriethoxysilane 98% (n-isomer ≥ 85%)
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06366-09
L06366-18
CAAAAL06366-09EA
75.68
CAD
CAAAAL06366-09
CAAAAL06366-18
Octadecyltriethoxysilane 98% (n-isomer ≥ 85%)
Octadecyltriethoxysilane
Purity: 98%
CAS No.: 7399-00-0
Molecular Formula: CH3(CH2)17Si(OCH2CH3)3
Formula Weight: 416.77
Flash Point: 195°(383°F)
Refractive Index: 1.4380
Boiling Point: 165-169°/0.2mm
Beilstein Reference: 7025163
MDL No.: MFCD00039875
Formula:
C₂₄H₅₂O₃Si Boiling Pt: 165…169 °C/0.2 mm Density: 0.870 Flash Pt: 195 °C(383 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039875 CAS Number: 7399-00-0 EINECS: 230-995-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...