2',6'-Dichloroacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06348-03
L06348-06
CAAAAL06348-03EA
102.7
CAD
CAAAAL06348-03
CAAAAL06348-06
2',6'-Dichloroacetophenone 98%
2',6'-Dichloroacetophenone
Purity: 98%
CAS No.: 2040-05-3
Molecular Formula: C8H6Cl2O
Formula Weight: 189.04
Melting Point: 41-44°
Boiling Point: 176-180°/83mm
MDL No.: MFCD00016339
Formula:
C₈H₆Cl₂O Boiling Pt: 176…180 °C/83 mm Melting Pt: 41…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016339 CAS Number: 2040-05-3 EINECS: 218-035-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...