4-(Aminomethyl)pyridine 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L06345-14
L06345-22
CAAAAL06345-14EA
79.63
CAD
CAAAAL06345-14
CAAAAL06345-22
4-(Aminomethyl)pyridine 97+%
4-(Aminomethyl)pyridine
Purity: 97+%
CAS No.: 3731-53-1
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 108°(226°F)
Refractive Index: 1.5515
Odor: Amine-like
Melting Point: -8°
Boiling Point: 230°
Beilstein Reference: 108057
UN No.: 2735
MDL No.: MFCD00006449
Formula:
C₆H₈N₂ Boiling Pt: 230 °C Melting Pt: –8 °C Density: 1.069 Flash Pt: 108 °C (226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006449 CAS Number: 3731-53-1 EINECS: 223-092-6 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...