Hexyl acetate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Acetic acid hexyl ester
L06342-AK
L06342-AU
CAAAAL06342-AKEA
52.14
CAD
CAAAAL06342-AK
CAAAAL06342-AU
Hexyl acetate 99%
Hexyl acetate
Purity: 99%
CAS No.: 142-92-7
Molecular Formula: C8H16O2
Formula Weight: 144.21
Flash Point: 43°(109°F)
Refractive Index: 1.4090
Melting Point: -80°
Boiling Point: 167-169°
Beilstein Reference: 1747138
UN No.: 3271
MDL No.: MFCD00009524
Formula:
C₈H₁₆O₂ MW: 144.21 g/mol Boiling Pt: 167…169 °C Melting Pt: –80 °C Density: 0.873 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009524 CAS Number: 142-92-7 EINECS: 205-572-7 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...