3-Phenyl-2-propyn-1-ol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06337-03
L06337-06
L06337-14
CAAAAL06337-03EA
38.71
CAD
CAAAAL06337-03
CAAAAL06337-06
CAAAAL06337-14
3-Phenyl-2-propyn-1-ol 97%
3-Phenyl-2-propyn-1-ol
Purity: 97%
CAS No.: 1504-58-1
Molecular Formula: C6H5C≡CCH2OH
Formula Weight: 132.16
Flash Point:
Formula:
C₉H₈O Boiling Pt: 129…130 °C/10 mm Density: 1.078 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040914 CAS Number: 1504-58-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...