1,9-Nonanediol 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
1,9-Dihydroxy nonane, Nonane-1,9-diol
L06336-14
L06336-22
CAAAAL06336-14EA
72.05
CAD
CAAAAL06336-14
CAAAAL06336-22
1,9-Nonanediol 97%
1,9-Nonanediol
Purity: 97%
CAS No.: 3937-56-2
Molecular Formula: HO(CH2)9OH
Formula Weight: 160.26
Flash Point: 162°(323°F)
Melting Point: 45-48°
Boiling Point: 288°
Beilstein Reference: 1737531
MDL No.: MFCD00002991
Formula:
C₉H₂₀O₂ Boiling Pt: 288 °C Melting Pt: 45…48 °C Density: 0.950 Flash Pt: 162 °C (323 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002991 CAS Number: 3937-56-2 EINECS: 223-517-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...