3-Ethyl-1-pentyne-3-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Ethylpent-1-yn-3-ol, 3-Ethyl-3-hydroxy-1-pentyne
L06335-06
L06335-14
CAAAAL06335-06EA
156.1
CAD
CAAAAL06335-06
CAAAAL06335-14
CAAAAL06335-22
3-Ethyl-1-pentyne-3-ol 98%
3-Ethyl-1-pentyne-3-ol
Purity: 98%
CAS No.: 6285-06-9
Molecular Formula: C7H12O
Formula Weight: 112.17
Flash Point: 38°(100°F)
Refractive Index: 1.4380
Boiling Point: 138-140°
Beilstein Reference: 1740944
UN No.: 1987
MDL No.: MFCD00014392
Formula:
C₇H₁₂O Boiling Pt: 138…140 °C Density: 0.873 Flash Pt: 38 °C (100 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014392 CAS Number: 6285-06-9 EINECS: 228-512-1 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...