L(+)-Cysteine hydrochloride, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06328-14
L06328-22
CAAAAL06328-14EA
94.17
CAD
CAAAAL06328-14
CAAAAL06328-22
L(+)-Cysteine hydrochloride, anhydrous 98%
L(+)-Cysteine hydrochloride
Purity: 98%
CAS No.: 52-89-1
Molecular Formula: C3H7NO2S.HCl
Formula Weight: 157.62
Odor: Irritating
pH: 0.8-1.2
Melting Point: ca 180° dec.
Beilstein Reference: 3560277
Merck Reference: 14,2781
Optical Rotation: +6.4° (c=5 in 5N HCl)
MDL No.: MFCD00064553
Formula:
HSCH₂CH(NH₂)COOH·HCl Melting Pt: ∼ 180 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064553 CAS Number: 52-89-1 EINECS: 200-157-7 Merck Index: 14,2781 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...