Hexyl benzoate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06293-14
L06293-22
CAAAAL06293-14EA
54.51
CAD
CAAAAL06293-14
CAAAAL06293-22
Hexyl benzoate 98+%
Hexyl benzoate
Purity: 98+%
CAS No.: 6789-88-4
Molecular Formula: C13H18O2
Formula Weight: 206.29
Flash Point: 148°(298°F)
Refractive Index: 1.4930
Boiling Point: 272°
Beilstein Reference: 2048117
MDL No.: MFCD00051714
Formula:
C₁₃H₁₈O₂ Boiling Pt: 272 °C Density: 0,980 Flash Pt: 148 °C(298 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051714 CAS Number: 6789-88-4 EINECS: 229-856-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...