4,5-Dichloroimidazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4,5-Dichloro-1H-imidazole
L06262-06
L06262-14
CAAAAL06262-06EA
91.17
CAD
CAAAAL06262-06
CAAAAL06262-14
4,5-Dichloroimidazole 98%
4,5-Dichloroimidazole
Purity: 98%
CAS No.: 15965-30-7
Molecular Formula: C3H2Cl2N2
Formula Weight: 136.97
Melting Point: 179-182°
Beilstein Reference: 606155
Formula:
C₃H₂Cl₂N₂ Melting Pt: 179…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005195 CAS Number: 15965-30-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...