Methyl isovalerate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Isopropyl acetic acid methyl ester, Isovaleric acid methyl ester, 3-Methylbutyric acid emthyl ester, Methyl isopentanoate, 3-Methylbutanoic acid methyl ester

L06241-18 L06241-30
CAAAAL06241-18EA 70.15 CAD
CAAAAL06241-18 CAAAAL06241-30
Methyl isovalerate 98%
Methyl isovalerate

Purity: 98% CAS No.: 556-24-1 Molecular Formula: (CH3)2CHCH2CO2CH3 Formula Weight: 116.17 Flash Point: 16°(60°F) Refractive Index: 1.3936 Boiling Point: 115-117° Beilstein Reference: 1699922 Merck Reference: 14,6093 UN No.: 2400 MDL No.: MFCD00042866

Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 115…117 °C
Density: 0.880
Flash Pt: 16 °C (60 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042866
CAS Number: 556-24-1
EINECS: 209-117-3
UN: 2400
ADR: 3 II
Merck Index: 14,6093

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR