2-(4-Methoxyphenyl)ethanol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Methoxyphenethyl alcohol

L06158-09 L06158-18
CAAAAL06158-09EA 67.94 CAD
CAAAAL06158-09 CAAAAL06158-18
2-(4-Methoxyphenyl)ethanol 98%
2-(4-Methoxyphenyl)ethanol

Purity: 98% CAS No.: 702-23-8 Molecular Formula: C9H12O2 Formula Weight: 152.19 Flash Point: 110°(230°F) Refractive Index: 1.5370 Melting Point: 26-30° Boiling Point: 334-336° Beilstein Reference: 2043563 MDL No.: MFCD00002900

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 334…336 °C
Melting Pt: 26…30 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002900
CAS Number: 702-23-8
EINECS: 211-866-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR