2,2'-Dichlorobenzophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2-DBP, Bis(2-chlorophenyl) ketone
L06144-03
L06144-06
CAAAAL06144-03EA
96.06
CAD
CAAAAL06144-03
CAAAAL06144-06
2,2'-Dichlorobenzophenone 98+%
2,2'-Dichlorobenzophenone
Purity: 98+%
CAS No.: 5293-97-0
Molecular Formula: C13H8Cl2O
Formula Weight: 251.11
Melting Point: 50-53°
Beilstein Reference: 2050630
Formula:
C₁₃H₈Cl₂O Melting Pt: 50…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039303 CAS Number: 5293-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...