2-Methyl-1-butene-3-yne 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Methylbut-1-en-3-yne
L06111-06
L06111-14
CAAAAL06111-06EA
100.49
CAD
CAAAAL06111-06
CAAAAL06111-14
2-Methyl-1-butene-3-yne 97%
2-Methyl-1-butene-3-yne
Purity: 97%
CAS No.: 78-80-8
Molecular Formula: C5H6
Formula Weight: 66.10
Storage Temperature: 2°C to 8°C
Flash Point: -45°(-49°F)
Refractive Index: 1.4180
Odor: Pungent
Melting Point: -113°
Boiling Point: 32-33°
Beilstein Reference: 969175
UN No.: 3295
MDL No.: MFCD00008599
Formula:
C₅H₆ Boiling Pt: 32…33 °C Melting Pt: –113 °C Density: 0.694 Flash Pt: -45 °C(-49 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008599 CAS Number: 78-80-8 EINECS: 201-144-9 UN: 3295 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...