trans-2-Pentenal 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trans-2-pentenal, trans-2-Pentenal, (E)-2-Pentenal
L06108-06
L06108-14
CAAAAL06108-06EA
69.05
CAD
CAAAAL06108-06
CAAAAL06108-14
trans-2-Pentenal 96%
trans-2-Pentenal
Purity: 96%
CAS No.: 1576-87-0
Molecular Formula: C5H8O
Formula Weight: 84.12
Flash Point: 22°(71°F)
Refractive Index: 1.4430
Odor: Fruit-like
Boiling Point: 125°
Beilstein Reference: 1719742
UN No.: 1989
MDL No.: MFCD00009615
Formula:
C₅H₈O MW: 84.12 g/mol Boiling Pt: 125 °C Density: 0.860 Flash Pt: 22 °C (71 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009615 CAS Number: 1576-87-0 EINECS: 216-414-1 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...