Molecular sieve 13X (1.0 nm, 10 Å), beads 3-5 mm (0.12-0.20 in)

Supplier: Thermo Scientific Chemicals

L06085-0B L06085-0I L06085-30
CAAAAL06085-0BEA 143.78 CAD
CAAAAL06085-0B CAAAAL06085-0I CAAAAL06085-30
Molecular sieve 13X (1.0 nm, 10 Å), beads 3-5 mm (0.12-0.20 in)
Molecular sieve 13X (1.0 nm, 10 Å)

CAS No.: 63231-69-6 MDL No.: MFCD00131613

Storage Temperature: Ambient
MDL Number: MFCD00164506
CAS Number: 63231-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR