3-Pentanol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L06065-22
L06065-36
CAAAAL06065-22EA
112.5
CAD
CAAAAL06065-22
CAAAAL06065-36
3-Pentanol 98+%
3-Pentanol
Purity: 98+%
CAS No.: 584-02-1
Molecular Formula: C5H12O
Formula Weight: 88.15
Flash Point: 33°(91°F)
Refractive Index: 1.4100
Odor: Characteristic
Melting Point: -8°
Boiling Point: 114-115°
Beilstein Reference: 1730964
Merck Reference: 14,7120
UN No.: 1105
MDL No.: MFCD00004574
Formula:
C₅H₁₂O Boiling Pt: 114…115 °C Melting Pt: –8 °C Density: 0.817 Flash Pt: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004574 CAS Number: 584-02-1 EINECS: 209-526-7 UN: 1105 ADR: 3,III Merck Index: 14,7120 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...