(±)-2-Aminobutyric acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-DL-Abu-OH, DL-2-Aminobutyric acid, DL-α-Amino-N-Butyric Acid

L06035-14 L06035-22
CAAAAL06035-14EA 70.63 CAD
CAAAAL06035-14 CAAAAL06035-22
(±)-2-Aminobutyric acid 99%
(±)-2-Aminobutyric acid

Purity: 99% CAS No.: 2835-81-6 Molecular Formula: C4H9NO2 Formula Weight: 103.12 Melting Point: ca 291° dec. Beilstein Reference: 635889 Merck Reference: 14,428 MDL No.: MFCD00008093

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Melting Pt: ca 291 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008093
CAS Number: 2835-81-6
EINECS: 220-616-5
Merck Index: 14,428

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR