5-Methyl-1-hexyne 98%

Supplier: Thermo Scientific Chemicals
Danger

L06014-06 L06014-14
CAAAAL06014-06EA 102.7 CAD
CAAAAL06014-06 CAAAAL06014-14
5-Methyl-1-hexyne 98%
5-Methyl-1-hexyne

Purity: 98% CAS No.: 2203-80-7 Molecular Formula: (CH3)2CH(CH2)2C≡CH Formula Weight: 96.17 Flash Point: -4°(24°F) Refractive Index: 1.4075 Boiling Point: 91-93° Beilstein Reference: 1731845 UN No.: 3295 MDL No.: MFCD00041610

Formula: C₇H₁₂
Boiling Pt: 91…93 °C
Melting Pt: –125 °C
Density: 0.732
Flash Pt: –4 °C (24 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041610
CAS Number: 2203-80-7
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR