Triethyl orthoacetate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1,1-Triethoxyethane

L06004-0F L06004-AE L06004-AP
CAAAAL06004-0FEA 619.36 CAD
CAAAAL06004-0F CAAAAL06004-AE CAAAAL06004-AP
Triethyl orthoacetate 98%
Triethyl orthoacetate

Purity: 98% CAS No.: 78-39-7 Molecular Formula: CH3C(OCH2CH3)3 Formula Weight: 162.23 Flash Point: 32°(89°F) Refractive Index: 1.3970 Boiling Point: 142-143° Beilstein Reference: 506201 UN No.: 1993 MDL No.: MFCD00009223

Formula: C₈H₁₈O₃
Boiling Pt: 142…143 °C
Density: 0.887
Flash Pt: 32 °C(89 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009223
CAS Number: 78-39-7
EINECS: 201-112-4
UN: 3272
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR