(±)-2-Hexanol 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(±)-Methyl butyl carbinol, (±)-Butyl methyl carbinol, (±)-2-Hydroxy hexane
L05993-14
L05993-22
CAAAAL05993-14EA
96.22
CAD
CAAAAL05993-14
CAAAAL05993-22
(±)-2-Hexanol 99%
(±)-2-Hexanol
Purity: 99%
CAS No.: 626-93-7
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 45°(113°F)
Refractive Index: 1.4140
Odor: Characteristic
Melting Point: -23°
Boiling Point: 137-139°
Beilstein Reference: 1718996
UN No.: 2282
MDL No.: MFCD00004585
Formula:
C₆H₁₄O MW: 102.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004585 CAS Number: 626-93-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...