(±)-4-Phenyl-2-butanol 98%

Supplier: Thermo Scientific Chemicals

L05970-06 L05970-14
CAAAAL05970-06EA 71.26 CAD
CAAAAL05970-06 CAAAAL05970-14
(±)-4-Phenyl-2-butanol 98%
(±)-4-Phenyl-2-butanol

Purity: 98% CAS No.: 2344-70-9 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 110°(230°F) Refractive Index: 1.5170 Boiling Point: 239° Beilstein Reference: 1908294 MDL No.: MFCD00044349

Formula: (C₆H₅)CH₂CH₂CH(OH)CH₃
Boiling Pt: 239 °C
Density: 0.980
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00044349
CAS Number: 2344-70-9
EINECS: 219-055-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR