DL-Tryptophan 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-DL-Trp-OH, (+/-)-2-Amino-3-(3-indolyl)propionic acid, 2-Amino-3-(1H-indol-3-yl)propanoic acid

L05936-06 L05936-14 L05936-22
CAAAAL05936-06EA 44.56 CAD
CAAAAL05936-06 CAAAAL05936-14 CAAAAL05936-22
DL-Tryptophan 99%
DL-Tryptophan

Purity: 99% CAS No.: 54-12-6 Molecular Formula: C11H12N2O2 Formula Weight: 204.23 Melting Point: ca 280° dec. Beilstein Reference: 86196

Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: ∼ 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064339
CAS Number: 54-12-6
EINECS: 200-194-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR