Margaric acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05928-06
L05928-14
CAAAAL05928-06EA
80.74
CAD
CAAAAL05928-06
CAAAAL05928-14
Margaric acid 98%
Margaric acid
Purity: 98%
CAS No.: 506-12-7
Molecular Formula: C17H34O2
Formula Weight: 270.46
Melting Point: 59-63°
Boiling Point: 227°/100mm
Merck Reference: 14,5751
MDL No.: MFCD00002751
Formula:
C₁₇H₃₄O₂ Boiling Pt: 227 °C/100 mm Melting Pt: 59…63 °C Density: 0.853 Storage Temperature: Ambient |
MDL Number:
MFCD00002751 CAS Number: 506-12-7 EINECS: 208-027-1 Merck Index: 14,5751 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...