Methyl-3,4,5-trihydroxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl gallate, Gallic acid methyl ester, Methyl 3,4,5-trihydroxybenzoate

L05926-18 L05926-30
CAAAAL05926-18EA 89.43 CAD
CAAAAL05926-18 CAAAAL05926-30
Methyl-3,4,5-trihydroxybenzoate 98%
Methyl 3,4,5-trihydroxybenzoate

Purity: 98% CAS No.: 99-24-1 Molecular Formula: C8H8O5 Formula Weight: 184.15 Melting Point: 198-203° Beilstein Reference: 2113180 Merck Reference: 14,4345 MDL No.: MFCD00002194

Formula: C₈H₈O₅
MW: 184.15 g/mol
Boiling Pt: 450 °C (1013 hPa)
Melting Pt: 198…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00002194
CAS Number: 99-24-1
EINECS: 202-741-7
Merck Index: 14,4345

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR