Decyltrichlorosilane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trichloro(decyl)silane

L05912-09 L05912-18
CAAAAL05912-09EA 57.04 CAD
CAAAAL05912-09 CAAAAL05912-18
Decyltrichlorosilane 97%
Decyltrichlorosilane

Purity: 97% CAS No.: 13829-21-5 Molecular Formula: C10H21Cl3Si Formula Weight: 275.72 Flash Point: 110°(230°F) Refractive Index: 1.4520 Boiling Point: 133-137°/5mm Beilstein Reference: 1749722 UN No.: 2987 MDL No.: MFCD00013600

Formula: C₁₀H₂₁Cl₃Si
Boiling Pt: 133…137 °C/5 mm
Density: 1.049
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013600
CAS Number: 13829-21-5
EINECS: 237-540-3
UN: 2987
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR