Ethylbenzene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phenylethane
L05908-0F
L05908-AE
L05908-AP
CAAAAL05908-0FEA
183.28
CAD
CAAAAL05908-0F
CAAAAL05908-AE
CAAAAL05908-AP
Ethylbenzene 99%
Ethylbenzene
Purity: 99%
CAS No.: 100-41-4
Molecular Formula: C8H10
Formula Weight: 106.17
Flash Point: 15°(59°F)
Refractive Index: 1.4950
Odor: Petrol-like
Melting Point: -95°
Boiling Point: 135-136°
Beilstein Reference: 1901871
Merck Reference: 14,3765
UN No.: 1175
MDL No.: MFCD00011647
Formula:
C₈H₁₀ MW: 106.17 g/mol Boiling Pt: 135…136 °C Melting Pt: –95 °C Density: 0.867 Flash Pt: 15 °C (59 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011647 CAS Number: 100-41-4 EINECS: 202-849-4 UN: 1175 ADR: 3,II Merck Index: 14,3765 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...