4,4'-Dimethylbenzhydrol 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Bis(4-methylphenyl)methanol, 4,4'-Dimethylbenzhydryl alcohol, Di-p-tolylmethanol, 4,4'-Dimethyl-α-phenylbenzyl alcohol, Bis(4-methylphenyl)carbinol, 4,4'-Dimethylphenyl phenyl carbinol, 4,4'-Dimethyldiphenylmethanol
L05904-03
L05904-06
CAAAAL05904-03EA
40.29
CAD
CAAAAL05904-03
CAAAAL05904-06
4,4'-Dimethylbenzhydrol 98%
4,4'-Dimethylbenzhydrol
Purity: 98%
CAS No.: 885-77-8
Molecular Formula: C15H16O
Formula Weight: 212.29
Melting Point: 69-73°
Beilstein Reference: 1874128
Formula:
C₁₅H₁₆O Melting Pt: 69…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017216 CAS Number: 885-77-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...