trans-2-Decenal 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05901-06
L05901-14
CAAAAL05901-06EA
96.22
CAD
CAAAAL05901-06
CAAAAL05901-14
trans-2-Decenal 95%
trans-2-Decenal
Purity: 95%
CAS No.: 3913-81-3
Molecular Formula: C10H18O
Formula Weight: 154.25
Storage Temperature: 2°C to 8°C
Flash Point: 93°(199°F)
Refractive Index: 1.4565
Boiling Point: 117°/17mm
Beilstein Reference: 1704445
MDL No.: MFCD00014679
Formula:
C₁₀H₁₈O Boiling Pt: 117 °C/17 mm Density: 0.840 Flash Pt: 93 °C (199 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00014679 CAS Number: 3913-81-3 EINECS: 223-474-2 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...