1-Propyl-4-iodobenzene 97%
Supplier: Thermo Scientific Chemicals
L05767-06
L05767-14
CAAAAL05767-06EA
118.97
CAD
CAAAAL05767-06
CAAAAL05767-14
1-Propyl-4-iodobenzene 97%
1-Propyl-4-iodobenzene
Purity: 97%
CAS No.: 126261-84-5
Molecular Formula: C9H11I
Formula Weight: 246.09
Refractive Index: 1.5790
Boiling Point: 240-242°
MDL No.: MFCD00051612
Formula:
C₉H₁₁I Boiling Pt: 240…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051612 CAS Number: 126261-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...