2-Phenylaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05728-03
L05728-06
CAAAAL05728-03EA
52.61
CAD
CAAAAL05728-03
CAAAAL05728-06
2-Phenylaniline 97%
2-Phenylaniline
Purity: 97%
CAS No.: 90-41-5
Molecular Formula: C12H11N
Formula Weight: 169.23
Flash Point: 153°(307°F)
Melting Point: 47-52°
Boiling Point: 299°
Beilstein Reference: 471874
MDL No.: MFCD00007701
Formula:
C₁₂H₁₁N Boiling Pt: 299 °C Melting Pt: 47…52 °C Flash Pt: 153 °C(307 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007701 CAS Number: 90-41-5 EINECS: 201-990-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...