Molecular sieve A5 (0.5 nm, 5 Å), beads 3-5 mm (0.12-0.20 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets

L05722-0B L05722-30
CAAAAL05722-0BEA 191.18 CAD
CAAAAL05722-0B CAAAAL05722-30
Molecular sieve A5 (0.5 nm, 5 Å), beads 3-5 mm (0.12-0.20 in)
Molecular sieve A5 (0.5 nm, 5 Å)

CAS No.: 69912-79-4 MDL No.: MFCD00131613

MDL Number: MFCD00131613
CAS Number: 69912-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR