trans-4-Decenal 96% may cont. ca 7% cis-isomer
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(E)-4-Decenal
L05715-06
L05715-14
CAAAAL05715-06EA
76.79
CAD
CAAAAL05715-06
CAAAAL05715-14
trans-4-Decenal 96% may cont. ca 7% cis-isomer
trans-4-Decenal
Purity: 96%
CAS No.: 65405-70-1
Molecular Formula: C10H18O
Formula Weight: 154.25
Storage Temperature: 2°C to 8°C
Flash Point: 85°(185°F)
Refractive Index: 1.4450
Boiling Point: 80°/4mm
Beilstein Reference: 4230058
MDL No.: MFCD00065447
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Boiling Pt: 80 °C/4 mm Density: 0.842 Flash Pt: 85 °C(185 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00065447 CAS Number: 65405-70-1 EINECS: 265-741-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...