n-Nonadecane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-nonadecane

L05701-14 L05701-22
CAAAAL05701-14EA 53.72 CAD
CAAAAL05701-14 CAAAAL05701-22
n-Nonadecane 99%
n-Nonadecane

Purity: 99% CAS No.: 629-92-5 Molecular Formula: CH3(CH2)17CH3 Formula Weight: 268.53 Flash Point: 168°(334°F) Refractive Index: 1.4410 Melting Point: 32-34° Boiling Point: 330-331° Beilstein Reference: 1742892 MDL No.: MFCD00009012

Formula: H₃C(CH₂)₁₇CH₃
MW: 268.53 g/mol
Boiling Pt: 330…331 °C
Melting Pt: 32…34 °C
Density: 0.786
Flash Pt: 168 °C (334 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009012
CAS Number: 629-92-5
EINECS: 211-116-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR