(±)-2-Decanol 98%
Supplier: Thermo Scientific Chemicals
Warning
|
L05688-06
L05688-14
CAAAAL05688-06EA
49.45
CAD
CAAAAL05688-06
CAAAAL05688-14
(±)-2-Decanol 98%
(±)-2-Decanol
Purity: 98%
CAS No.: 1120-06-5
Molecular Formula: C10H22O
Formula Weight: 158.29
Flash Point: 85°(185°F)
Refractive Index: 1.4340
Melting Point: -5°
Boiling Point: 211°
Beilstein Reference: 1719664
MDL No.: MFCD00004594
Formula:
C₁₀H₂₂O Boiling Pt: 211 °C Melting Pt: –5 °C Density: 0.825 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004594 CAS Number: 1120-06-5 EINECS: 214-296-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...