Sodium methoxide 98%

Supplier: Thermo Scientific Chemicals
Danger

L05673-0P L05673-22 L05673-36
CAAAAL05673-0PEA 235.42 CAD
CAAAAL05673-0P CAAAAL05673-22 CAAAAL05673-36
Sodium methoxide 98%
Sodium methoxide

Purity: 98% CAS No.: 124-41-4 Molecular Formula: CH3NaO Formula Weight: 54.02 Melting Point: 300° Beilstein Reference: 3592982 Merck Reference: 14,8643 UN No.: 1431 MDL No.: MFCD00012179

Formula: CH₃NaO
Melting Pt: >300 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012179
CAS Number: 124-41-4
EINECS: 204-699-5
UN: 1431
ADR: 4.2,II
Merck Index: 14,8643

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR