N-Ethylbenzylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L05651-AC
L05651-AE
L05651-AP
CAAAAL05651-ACEA
58.93
CAD
CAAAAL05651-AC
CAAAAL05651-AE
CAAAAL05651-AP
N-Ethylbenzylamine 97%
N-Ethylbenzylamine
Purity: 97%
CAS No.: 14321-27-8
Molecular Formula: C6H5CH2NHCH2CH3
Formula Weight: 135.21
Flash Point: 66°(150°F)
Refractive Index: 1.5110
Boiling Point: 191-194°
Beilstein Reference: 386023
UN No.: 2735
MDL No.: MFCD00009031
Formula:
C₆H₅CH₂NHCH₂CH₃ Boiling Pt: 191…194 °C Density: 0.910 Flash Pt: 66 °C (150 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009031 CAS Number: 14321-27-8 EINECS: 238-265-1 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...