2-(2-Aminoethyl)pyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Pyridylethylamine, 2-(2-Pyridyl)ethylamine, 2-Pyridin-2-ylethanamine
L05627-03
L05627-06
L05627-14
CAAAAL05627-03EA
49.93
CAD
CAAAAL05627-03
CAAAAL05627-06
CAAAAL05627-14
2-(2-Aminoethyl)pyridine 98%
2-(2-Aminoethyl)pyridine
Purity: 98%
CAS No.: 2706-56-1
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 100°(212°F)
Refractive Index: 1.5360
Odor: Amine-like
Boiling Point: 208°
Beilstein Reference: 111208
UN No.: 2735
MDL No.: MFCD00006367
Formula:
C₇H₁₀N₂ Boiling Pt: 208 °C Density: 1.020 Flash Pt: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006367 CAS Number: 2706-56-1 EINECS: 220-295-1 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...