2-(4-Fluorophenyl)ethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Fluorophenethyl alcohol

L05618-06 L05618-14
CAAAAL05618-06EA 104.6 CAD
CAAAAL05618-06 CAAAAL05618-14
2-(4-Fluorophenyl)ethanol 97%
2-(4-Fluorophenyl)ethanol

Purity: 97% CAS No.: 7589-27-7 Molecular Formula: C8H9FO Formula Weight: 140.16 Flash Point: 104°(219°F) Refractive Index: 1.5065 Boiling Point: 118°/20mm Beilstein Reference: 2354310 MDL No.: MFCD00002898

Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 118 °C/20 mm
Density: 1.121
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002898
CAS Number: 7589-27-7
EINECS: 231-499-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR