2-Octyn-1-ol 98%

Supplier: Thermo Scientific Chemicals
Warning

L05596-09 L05596-18
CAAAAL05596-09EA 110.28 CAD
CAAAAL05596-09 CAAAAL05596-18
2-Octyn-1-ol 98%
2-Octyn-1-ol

Purity: 98% CAS No.: 20739-58-6 Molecular Formula: CH3(CH2)4C≡CCH2OH Formula Weight: 126.20 Flash Point: 91°(195°F) Refractive Index: 1.4560 Boiling Point: 76-78°/2mm Beilstein Reference: 1744120 MDL No.: MFCD00039542

Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 76…78 °C/2 mm
Density: 0.880
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039542
CAS Number: 20739-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR